4,5-Dihydro-2-(methylthio)-1H-imidazole-1-carboxylic acid methyl ester 95%
Leverandør: Apollo Scientific
Synonymer: Methyl 2-(methylthio)-4,5-dihydro-1H-imidazole-1-carboxylate
OR944682-1G OR944682-250MG
APOSOR944682-1GEA 1900 NOK
APOSOR944682-1G APOSOR944682-250MG
4,5-Dihydro-2-(methylthio)-1H-imidazole-1-carboxylic acid methyl ester 95%
4,5-Dihydro-2-(methylthio)-1H-imidazole-1-carboxylic acid methyl ester
Formel: C₆H₁₀N₂O₂S
MW: 174,221 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD25965248
CAS-nummer: 60546-77-2