4-Methyl-2-nitrobenzylalcohol 95%

Leverandør: Apollo Scientific

Synonymer: (4-Methyl-2-nitrophenyl)methanol

OR947345-1G OR947345-5G
APOSOR947345-1GEA 1350 NOK
APOSOR947345-1G APOSOR947345-5G
4-Methyl-2-nitrobenzylalcohol 95%
4-Methyl-2-nitrobenzylalcohol
Formel: C₈H₉NO₃
MW: 167,162 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08275452
CAS-nummer: 22996-24-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR