3-Amino-5-bromopyridine-2-carbonitrile 98%
Leverandør: Apollo Scientific
Synonymer:
3-Amino-5-bromopicolinonitrile
OR959411-5G
APOSOR959411-5GEA
1850
NOK
APOSOR959411-5G
3-Amino-5-bromopyridine-2-carbonitrile 98%
3-Amino-5-bromopyridine-2-carbonitrile
Formel:
C₆H₄BrN₃ MW: 198,02 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD07375025 CAS-nummer: 573675-27-1 FN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...