2-Ethylsulphanyl-1H-benzoimidazole 95%

Leverandør: Apollo Scientific

OR963937-1G
APOSOR963937-1GEA 0 NOK
APOSOR963937-1G
2-Ethylsulphanyl-1H-benzoimidazole 95%
2-Ethylsulphanyl-1H-benzoimidazole
Formel: C₉H₁₀N₂S
MW: 178,254 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00223154
CAS-nummer: 14610-11-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR