Methyl 2-cyano-5-fluorobenzoate 95+%

Leverandør: Apollo Scientific

PC201268-100MG PC201268-250MG PC201268-1G
APOSPC201268-100MGEA 1075 NOK
APOSPC201268-100MG APOSPC201268-250MG APOSPC201268-1G
Methyl 2-cyano-5-fluorobenzoate 95+%
Methyl 2-cyano-5-fluorobenzoate
Formel: C₉H₆FNO₂
MW: 179,148 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11042789
CAS-nummer: 606080-43-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR