2-Bromo-4-fluoro-5-methylaniline 97%
Leverandør: Apollo Scientific
|
Warning
|
PC3785-5G
APOSPC3785-5GEA
1075
NOK
APOSPC3785-5G
2-Bromo-4-fluoro-5-methylaniline 97%
2-Bromo-4-fluoro-5-methylaniline
Formel:
C₇H₇BrFN MW: 204,04 g/mol Smeltepunkt: 65…67 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD11042843 CAS-nummer: 1065076-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...