4-Bromo-2-fluorophenyl trifluoromethanesulphonate
Leverandør: Apollo Scientific
Warning
PC500046-1G PC500046-5G PC500046-250MG
APOSPC500046-1GEA 1725 NOK
APOSPC500046-1G APOSPC500046-5G APOSPC500046-250MG
4-Bromo-2-fluorophenyl trifluoromethanesulphonate
4-Bromo-2-fluorophenyl trifluoromethanesulphonate
Formel: C₇H₃BrF₄O₃S
MW: 323,06 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD25541808
CAS-nummer: 872054-03-0