2-Amino-1-(4-fluoro-3-methyl-phenyl)-ethanone monohydrochloride 95%
Leverandør: Apollo Scientific
PC520813-1G
PC520813-250MG
PC520813-500MG
APOSPC520813-1GEA
7175
NOK
APOSPC520813-1G
APOSPC520813-250MG
APOSPC520813-500MG
2-Amino-1-(4-fluoro-3-methyl-phenyl)-ethanone monohydrochloride 95%
2-Amino-1-(4-fluoro-3-methyl-phenyl)-ethanone monohydrochloride
Formel:
C₉H₁₀FNO·ClH MW: 203,641 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD11226358 CAS-nummer: 1177342-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...