5-Fluoro-6-methoxy-1H-indazole 95+%

Leverandør: Apollo Scientific
Warning

PC535075-100MG PC535075-250MG
APOSPC535075-100MGEA 1325 NOK
APOSPC535075-100MG APOSPC535075-250MG
5-Fluoro-6-methoxy-1H-indazole 95+%
5-Fluoro-6-methoxy-1H-indazole
Formel: C₈H₇FN₂O
MW: 166,152 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD11007884
CAS-nummer: 1082041-57-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR