2-Bromoethyl trifluoromethanesulphonate 95%

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Bromoethyl triflate

PC57000-1G PC57000-5G
APOSPC57000-1GEA 3325 NOK
APOSPC57000-1G APOSPC57000-5G
2-Bromoethyl trifluoromethanesulphonate 95%
2-Bromoethyl trifluoromethanesulphonate
Formel: C₃H₄BrF₃O₃S
MW: 257,026 g/mol
Kokepunkt: 230 °C
Tetthet: 1,9 g/cm³ (25 °C)
Flash punkt: >110 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD11656439
CAS-nummer: 103935-47-3
FN: 2927
ADR: 6.1+8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR