2,5-Difluorostyrene 99%

Leverandør: Apollo Scientific
Warning

Synonymer: 1,4-Difluoro-2-vinylbenzene

PC6275-1G PC6275-5G
APOSPC6275-1GEA 2875 NOK
APOSPC6275-1G APOSPC6275-5G
2,5-Difluorostyrene 99%
2,5-Difluorostyrene
Formel: C₈H₆F₂
MW: 140,13 g/mol
Kokepunkt: 56 °C (30 mm Hg)
Tetthet: 1,1306 g/cm³ (25 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00142841
CAS-nummer: 398-70-9
FN: 1993
ADR: 3,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR