2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile 97%
Leverandør: Apollo Scientific
PC905878-5G
APOSPC905878-5GEA
1050
NOK
APOSPC905878-5G
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile 97%
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile
Formel:
C₁₆H₁₀FN₃O MW: 279,269 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD28133450 CAS-nummer: 1021298-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...