N-Acetylglycine(Ac-Gly-OH)
Leverandør: Apollo Scientific
Warning
Synonymer: Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid
BIA1745-100G BIA1745-500G
APOSBIA1745-100GEA 340 NOK
APOSBIA1745-100G APOSBIA1745-500G
N-Acetylglycine(Ac-Gly-OH)
N-Acetylglycine(Ac-Gly-OH)
Formel: C₄H₇NO₃
MW: 117.1 g/mol
Smeltepunkt: 207…209 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00004275
CAS-nummer: 543-24-8
EINECS: 208-839-6