10,11-Dehydrocurvularin (from Penicillium citrinum)
Leverandør: Apollo Scientific
|
Warning
|
BIA4323-1MG
BIA4323-5MG
APOSBIA4323-1MGEA
1175
NOK
APOSBIA4323-1MG
APOSBIA4323-5MG
10,11-Dehydrocurvularin (from Penicillium citrinum)
10,11-Dehydrocurvularin
Smeltepunkt:
200 °C Lagringstemperatur: Fryseskap |
CAS-nummer:
1095588-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...