Chenodeoxycholic acid
Leverandør: Apollo Scientific
Warning
APOSBIB6026-100GEA 2525 NOK
APOSBIB6026-100G APOSBIB6026-50G APOSBIB6026-25G
Chenodeoxycholic acid
Chenodeoxycholic acid
Bile acid.
Formel: C₂₄H₄₀O₄
MW: 392,58 g/mol
Smeltepunkt: 162…168 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064142
CAS-nummer: 474-25-9
EINECS: 207-481-8