SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)
Leverandør: Apollo Scientific
Warning
Synonymer: 2,5-Dioxopyrrolidin-1-yl 3-(pyridin-2-yldisulphanyl)propanoate, 3-(2-Pyridyldithio)propionic Acid N-Succinimidyl Ester, SPDP, 3-(2-Pyridyldithio)-Propionic Acid N-Hydroxysuccinimide Ester
BICL210-25MG
APOSBICL210-25MGEA 1275 NOK
APOSBICL210-25MG
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)
Primary amine & Sulphhydryl reactiveThiolation -CleavableRef: Cumber,J.Aet al(1985)Methods Enzymol.112, 207-224
Formel: C₁₂H₁₂N₂O₄S₂
MW: 312,36 g/mol
Smeltepunkt: 84…86 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00009636
CAS-nummer: 68181-17-9
EINECS: 269-034-3