(2-Hydroxylpropyl)-γ-cyclodextrine (DS~4.5)

Leverandør: Apollo Scientific

Synonymer: (2-Hydroxypropyl)-γ-Cyclodextrin, HGC

BICY1008-25G BICY1008-100G
APOSBICY1008-25GEA 4125 NOK
APOSBICY1008-25G APOSBICY1008-100G
(2-Hydroxylpropyl)-γ-cyclodextrine (DS~4.5)
(2-Hydroxylpropyl)-γ-cyclodextrine
Formel: C₇₂H₁₂₈O₄₈
MW: 1761,7600 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00148890
CAS-nummer: 128446-34-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR