L(+)-Glutamin ≥99%
Leverandør: Apollo Scientific
Warning
Synonymer: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
BIG0708-100G BIG0708-1KG BIG0708-500G
APOSBIG0708-100GEA 270 NOK
APOSBIG0708-100G APOSBIG0708-1KG APOSBIG0708-500G
L(+)-Glutamin ≥99%
L(+)-Glutamin
Pyrogen free
Formel: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Kokepunkt: 355
Smeltepunkt: 185 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008044
CAS-nummer: 56-85-9
EINECS: 200-292-1
Merck Indeks: 13,04484