Patulin (from Penicillium expansum)
Leverandør: Apollo Scientific
Danger
BIP1003-10MG BIP1003-5MG
APOSBIP1003-10MGEA 2425 NOK
APOSBIP1003-10MG APOSBIP1003-5MG
Patulin (from Penicillium expansum)
Patulin
Mycotoxin
Formel: C₇H₆O₄
Smeltepunkt: 109…111 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00005858
CAS-nummer: 149-29-1
EINECS: 205-735-2
FN: 3462
ADR: 6.1,II