Aluminium nitrate hydrate 99,9%
Leverandør: Apollo Scientific
Warning
IN1024-1KG IN1024-200G
APOSIN1024-1KGEA 3150 NOK
APOSIN1024-1KG APOSIN1024-200G
Aluminium nitrate hydrate 99,9%
Aluminium nitrate hydrate
Formel: AlN₃O₉·H₂O
MW: 231,01 g/mol
Smeltepunkt: 73,5 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003419
CAS-nummer: 25838-59-9
FN: 1438
ADR: 5.1,III