Dibismuth molybdenum hexaoxide 99%
Leverandør: Apollo Scientific
|
Warning
|
Synonymer:
Dibismuth(III) molybdenum(VI) oxide
IN1246-250G
IN1246-50G
IN1246-1KG
APOSIN1246-250GEA
5275
NOK
APOSIN1246-250G
APOSIN1246-50G
APOSIN1246-1KG
Dibismuth molybdenum hexaoxide 99%
Dibismuth molybdenum hexaoxide
Formel:
Bi₂MoO₆ Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00064779 CAS-nummer: 13565-96-3 EINECS: 236-955-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...