Ruthenium(III) chloride hydrate 97%
Leverandør: Apollo Scientific
Danger
Synonymer: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate
IN3062-5G IN3062-1G IN3062-25G
APOSIN3062-5GEA 1700 NOK
APOSIN3062-5G APOSIN3062-1G APOSIN3062-25G
Ruthenium(III) chloride hydrate 97%
Ruthenium(III) chloride hydrate
Formel: Cl₃Ru·xH₂O
MW: 207,43 g/mol (anhy)
Smeltepunkt: 100 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149844
CAS-nummer: 14898-67-0
EINECS: 233-167-5
FN: 3260
ADR: 8,II