5-tert-Butyl-2-methyl-3-furoic acid

Leverandør: Apollo Scientific
Warning

OR01668-1G OR01668-250MG
APOSOR01668-1GEA 1075 NOK
APOSOR01668-1G APOSOR01668-250MG
5-tert-Butyl-2-methyl-3-furoic acid
5-tert-Butyl-2-methyl-3-furoic acid
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Smeltepunkt: 89…92 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00221068
CAS-nummer: 38422-62-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR