4-Methoxysalicylaldehyde 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Hydroxy-4-methoxybenzaldehyde

OR0913-100G OR0913-25G OR0913-500G
APOSOR0913-100GEA 1475 NOK
APOSOR0913-100G APOSOR0913-25G APOSOR0913-500G
4-Methoxysalicylaldehyde 97%
4-Methoxysalicylaldehyde
Formel: C₈H₈O₃
MW: 152,15 g/mol
Smeltepunkt: 40…43 °C
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003327
CAS-nummer: 673-22-3
EINECS: 211-604-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR