(1-Methyl-1H-imidazol-5-yl)methanol

Leverandør: Apollo Scientific
Warning

Synonymer: 5-(Hydroxymethyl)-1-methyl-1H-imidazole

OR10499-5G
APOSOR10499-5GEA 1850 NOK
APOSOR10499-5G
(1-Methyl-1H-imidazol-5-yl)methanol
(1-Methyl-1H-imidazol-5-yl)methanol
Formel: C₅H₈N₂O
MW: 112,1298 g/mol
Smeltepunkt: 110…112 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02179522
CAS-nummer: 38993-84-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR