3-(Methylsulphonyl)phenylboronic acid 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 3-(Methanesulphonyl)phenylboronic acid, 3-(Methylsulphonyl)benzeneboronic acid, (3-(Methylsulphonyl)phenyl)boronic acid

OR10565-1G OR10565-5G
APOSOR10565-1GEA 290 NOK
APOSOR10565-1G APOSOR10565-5G
3-(Methylsulphonyl)phenylboronic acid 98%
3-(Methylsulphonyl)phenylboronic acid
Formel: C₇H₉BO₄S
MW: 200,02 g/mol
Smeltepunkt: 102…110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03092935
CAS-nummer: 373384-18-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR