3-Nitrophenylboronic acid 97%

Leverandør: Apollo Scientific
Warning

Synonymer: (3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid

OR10571-5G
APOSOR10571-5GEA 270 NOK
APOSOR10571-5G
3-Nitrophenylboronic acid 97%
3-Nitrophenylboronic acid
Formel: C₆H₆BNO₄
MW: 166,93 g/mol
Smeltepunkt: 284…285 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007193
CAS-nummer: 13331-27-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR