Methylenediphosphonic acid 98%

Leverandør: Apollo Scientific
Danger

Synonymer: Medronic acid

OR10789-1G OR10789-5G
APOSOR10789-1GEA 340 NOK
APOSOR10789-1G APOSOR10789-5G
Methylenediphosphonic acid 98%
Methylenediphosphonic acid
Formel: CH₆O₆P₂
MW: 176 g/mol
Smeltepunkt: 197…204 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00002139
CAS-nummer: 1984-15-2
EINECS: 217-851-0
FN: 3261
ADR: 8,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR