3-(Ethoxycarbonyl)phenylboronic acid pinacol ester 98%

Leverandør: Apollo Scientific
Warning

Synonymer: Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Ethoxycarbonylphenylboronic acid, pinacol ester, Ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

OR10988-25G
APOSOR10988-25GEA 1725 NOK
APOSOR10988-25G
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester 98%
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Formel: C₁₅H₂₁BO₄
MW: 276,136 g/mol
Smeltepunkt: 40…44 °C
Flash punkt: 110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03093896
CAS-nummer: 269410-00-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR