6-Amino-5-chloronicotinic acid, Technical Grade
Leverandør: Apollo Scientific
Warning
OR110434-1G
APOSOR110434-1GEA 3050 NOK
APOSOR110434-1G
6-Amino-5-chloronicotinic acid, Technical Grade
6-Amino-5-chloronicotinic acid
Formel: C₆H₅ClN₂O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11132184
CAS-nummer: 41668-11-5