Ethyl-1H-indazole-6-carboxylate

Leverandør: Apollo Scientific
Warning

Synonymer: Ethyl 1H-indazole-6-carboxylate

OR111116-1G OR111116-5G OR111116-10G
APOSOR111116-1GEA 360 NOK
APOSOR111116-1G APOSOR111116-5G APOSOR111116-10G
Ethyl-1H-indazole-6-carboxylate
Ethyl-1H-indazole-6-carboxylate
Formel: C₁₀H₁₀N₂O₂
MW: 190,2000 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07371613
CAS-nummer: 713-09-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR