Methyl-3,5-dimethoxybenzoate 97%
Leverandør: Apollo Scientific
Synonymer:
Methyl 3,5-dimethoxybenzoate, 3,5-Dimethoxybenzoic acid methyl ester
OR11318-500G
APOSOR11318-500GEA
3075
NOK
APOSOR11318-500G
Methyl-3,5-dimethoxybenzoate 97%
Methyl-3,5-dimethoxybenzoate
Formel:
C₁₀H₁₂O₄ MW: 196,2 g/mol Kokepunkt: 297…299 °C Smeltepunkt: 42…43 °C Flash punkt: >110 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00008432 CAS-nummer: 2150-37-0 EINECS: 218-423-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...