2-Bromo-6-nitroaniline 99%

Leverandør: Apollo Scientific
Warning

OR11720-10G OR11720-1G OR11720-5G
APOSOR11720-10GEA 1425 NOK
APOSOR11720-10G APOSOR11720-1G APOSOR11720-5G
2-Bromo-6-nitroaniline 99%
2-Bromo-6-nitroaniline
Formel: C₆H₅BrN₂O₂
Smeltepunkt: 72…74 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09263476
CAS-nummer: 59255-95-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR