2-(Methylamino)nicotinonitrile

Leverandør: Apollo Scientific
Warning

OR12119-1G
APOSOR12119-1GEA 645 NOK
APOSOR12119-1G
2-(Methylamino)nicotinonitrile
2-(Methylamino)nicotinonitrile
Formel: C₇H₇N₃
MW: 133,1506 g/mol
Smeltepunkt: 85…87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08689752
CAS-nummer: 52583-87-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR