3-(4-Isobutylphenyl)acrylic acid

Leverandør: Apollo Scientific
Warning

Synonymer: 4-Isobutylcinnamic acid

OR13177-1G OR13177-250MG
APOSOR13177-1GEA 305 NOK
APOSOR13177-1G APOSOR13177-250MG
3-(4-Isobutylphenyl)acrylic acid
3-(4-Isobutylphenyl)acrylic acid
Formel: C₁₃H₁₆O₂
MW: 204,27 g/mol
Smeltepunkt: 168…169 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00800640
CAS-nummer: 66734-95-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR