4-[4-(4-Aminobenzoyl)piperazino]benzenecarbonitrile

Leverandør: Apollo Scientific
Warning

Synonymer: 4-[4-(4-Aminobenzoyl)piperazin-1-yl]benzonitrile

OR14359-1G OR14359-500MG
APOSOR14359-1GEA 4575 NOK
APOSOR14359-1G APOSOR14359-500MG
4-[4-(4-Aminobenzoyl)piperazino]benzenecarbonitrile
4-[4-(4-Aminobenzoyl)piperazino]benzenecarbonitrile
Formel: C₁₈H₁₈N₄O
MW: 306,37 g/mol
Smeltepunkt: 195 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06659560
CAS-nummer: 885949-73-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR