Benzyltrimethylammonium bromide
Leverandør: Apollo Scientific
|
Warning
|
APOSOR15010-25GEA
180
NOK
APOSOR15010-25G
Benzyltrimethylammonium bromide
Benzyltrimethylammonium bromide
Formel:
C₁₀H₁₆BrN MW: 230,15 g/mol Smeltepunkt: 228…232 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00011780 CAS-nummer: 5350-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...