3-Bromophenol 98%

Leverandør: Apollo Scientific
Warning

OR18331-100G OR18331-500G
APOSOR18331-100GEA 645 NOK
APOSOR18331-100G APOSOR18331-500G
3-Bromophenol 98%
3-Bromophenol
Formel: C₆H₅BrO
MW: 173,01 g/mol
Kokepunkt: 236 °C
Smeltepunkt: 28…32 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002253
CAS-nummer: 591-20-8
EINECS: 209-706-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR