Trimethylsilylacetylene 97%
Leverandør: Apollo Scientific
|
Danger
|
Synonymer:
Ethynyltrimethylsilane
OR18355-100G
OR18355-25G
OR18355-500G
APOSOR18355-100GEA
860
NOK
APOSOR18355-100G
APOSOR18355-25G
APOSOR18355-500G
Trimethylsilylacetylene 97%
Trimethylsilylacetylene
Formel:
C₅H₁₀Si MW: 98,22 g/mol Kokepunkt: 52…53 °C Tetthet: 0,71 g/cm³ (25 °C) Flash punkt: –34 °C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00008569 CAS-nummer: 1066-54-2 EINECS: 213-919-9 FN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...