Boc-L-pyroglutamic acid methyl ester
Leverandør: Apollo Scientific
Warning
Synonymer: Methyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, Methyl N-Boc-L-pyroglutamate, Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate, Boc-Pyr-OMe, (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-Boc-L-pyroglutamic acid methyll ester, (S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-(tert-Butoxycarbonyl)-L-pyroglutamic acid methyll ester, 1-(tert-Butyl) 2-methyl (S)-5-oxopyrrolidine-1,2-dicarboxylate, Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
OR18848-500G
APOSOR18848-500GEA 1150 NOK
APOSOR18848-500G
Boc-L-pyroglutamic acid methyl ester
Boc-L-pyroglutamic acid methyl ester
Formel: C₁₁H₁₇NO₅
MW: 243,26 g/mol
Smeltepunkt: 68…74 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06809720
CAS-nummer: 108963-96-8
FN: 3077
ADR: 9+EHS,III