Anthranilohydrazide
Leverandør: Apollo Scientific
Warning
Synonymer: 2-Aminobenzoic hydrazide, 2-Aminobenzoic acid hydrazide, 2-Aminobenzohydrazide, 2-Aminobenzoylhydrazide, Anthraniloyl Hydrazine, 2-Aminobenzene-1-carbohydrazide, 2-Aminobenzhydrazide
OR1896-100G OR1896-25G
APOSOR1896-100GEA 1225 NOK
APOSOR1896-100G APOSOR1896-25G
Anthranilohydrazide
Anthranilohydrazide
Formel: C₇H₉N₃O
MW: 151,17 g/mol
Smeltepunkt: 121…125 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017056
CAS-nummer: 1904-58-1