4-Isopropyl-6-(methylthio)-1,3,5-triazin-2-amine

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Amino-4-isopropyl-6-(methylthio)-1,3,5-triazine

OR27306-1G
APOSOR27306-1GEA 1125 NOK
APOSOR27306-1G
4-Isopropyl-6-(methylthio)-1,3,5-triazin-2-amine
4-Isopropyl-6-(methylthio)-1,3,5-triazin-2-amine
Formel: C₇H₁₂N₄S
MW: 184,26 g/mol
Smeltepunkt: 127…130 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00052790
CAS-nummer: 175204-55-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR