4-Iodobutyl acetate 96%

Leverandør: Apollo Scientific
Warning

OR27557-10G OR27557-50G
APOSOR27557-10GEA 735 NOK
APOSOR27557-10G APOSOR27557-50G
4-Iodobutyl acetate 96%
4-Iodobutyl acetate
Formel: C₆H₁₁IO₂
MW: 242,06 g/mol
Kokepunkt: 94…95 °C/5,5mmHg
Tetthet: 1,61 g/cm³ (25 °C)
Flash punkt: 91 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010664
CAS-nummer: 40596-44-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR