1-Carbamoylcyclopropane-1-carboxylic acid

Leverandør: Apollo Scientific
Warning

Synonymer: 1-Carbamoylcyclopropanecarboxylic acid

OR303011-1G
APOSOR303011-1GEA 325 NOK
APOSOR303011-1G
1-Carbamoylcyclopropane-1-carboxylic acid
1-Carbamoylcyclopropane-1-carboxylic acid
Formel: C₅H₇NO₃
MW: 129.12 g/mol
Smeltepunkt: 185 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00190650
CAS-nummer: 6914-74-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR