6-Bromo-3,4-dihydro-1H-quinolin-2-one

Leverandør: Apollo Scientific
Warning

OR30464-25G OR30464-5G
APOSOR30464-25GEA 1750 NOK
APOSOR30464-25G APOSOR30464-5G
6-Bromo-3,4-dihydro-1H-quinolin-2-one
6-Bromo-3,4-dihydro-1H-quinolin-2-one
Formel: C₉H₈BrNO
MW: 226,07 g/mol
Smeltepunkt: 170…172 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03839832
CAS-nummer: 3279-90-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR