2-Nitro-p-toluic acid 97%

Leverandør: Apollo Scientific
Warning

OR30706-25G
APOSOR30706-25GEA 2900 NOK
APOSOR30706-25G
2-Nitro-p-toluic acid 97%
2-Nitro-p-toluic acid
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Smeltepunkt: 160…164 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00033899
CAS-nummer: 27329-27-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR