1-(4-Chlorobenzoyl)-4-piperidinecarboxylic acid 95%

Leverandør: Apollo Scientific

OR33302-1G OR33302-5G
APOSOR33302-1GEA 2225 NOK
APOSOR33302-1G APOSOR33302-5G
1-(4-Chlorobenzoyl)-4-piperidinecarboxylic acid 95%
1-(4-Chlorobenzoyl)-4-piperidinecarboxylic acid
Formel: C₁₃H₁₄ClNO₃
MW: 267,71 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03014156
CAS-nummer: 379724-54-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR