8-(Benzyloxy)imidazo[1,2-a]pyridine 95%

Leverandør: Apollo Scientific

OR33317-1G OR33317-5G
APOSOR33317-1GEA 4150 NOK
APOSOR33317-1G APOSOR33317-5G
8-(Benzyloxy)imidazo[1,2-a]pyridine 95%
8-(Benzyloxy)imidazo[1,2-a]pyridine
Formel: C₁₄H₁₂N₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06739199
CAS-nummer: 96428-16-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR