3-Bromo-2-(2-bromoacetyl)thiophene 95%

Leverandør: Apollo Scientific

Synonymer: 2-Bromo-1-(3-bromothiophen-2-yl)ethan-1-one

OR33358-1G OR33358-5G
APOSOR33358-1GEA 4150 NOK
APOSOR33358-1G APOSOR33358-5G
3-Bromo-2-(2-bromoacetyl)thiophene 95%
3-Bromo-2-(2-bromoacetyl)thiophene
Formel: C₆H₄Br₂OS
Lagringstemperatur: Romtemperatur
CAS-nummer: 81216-95-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR