3-Bromo-2-iodoaniline
Leverandør: Apollo Scientific
|
Danger
|
OR400863-1G
OR400863-250MG
OR400863-5G
APOSOR400863-1GEA
1575
NOK
APOSOR400863-1G
APOSOR400863-250MG
APOSOR400863-5G
3-Bromo-2-iodoaniline
3-Bromo-2-iodoaniline
Formel:
C₆H₅BrIN MW: 297.9191 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD16622211 CAS-nummer: 150908-00-2 FN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...